Product Details
- SNP ID
-
rs2600714
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:174809841 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TCTGGGGTAAAGCTGATGAGATTTT[A/G]TTTGTTTTGACCATGGATCTCTCAC
- Phenotype
-
MIM: 118423
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CHN1
PubMed Links
Gene Details
- Gene
- CHN1
- Gene Name
- chimerin 1
View Full Product Details