Product Details
- SNP ID
-
rs2591030
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:61067006 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTGAGCACGTGCTGTGTGCACAGTC[A/C]TGAGTCAGGAGCCATAAATGGGGGG
- Phenotype
-
MIM: 611292
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
CLVS1
PubMed Links
Gene Details
- Gene
- CLVS1
- Gene Name
- clavesin 1
View Full Product Details