Product Details
- SNP ID
-
rs2257505
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:56911296 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTTCAATAAACAATCAGAAATCTTG[A/T]CTATGCTTCCAGAATCTGTTAAATC
- Phenotype
-
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
MIER3
PubMed Links
Gene Details
- Gene
- MIER3
- Gene Name
- MIER family member 3
There are no transcripts associated with this gene.
- Gene
- SETD9
- Gene Name
- SET domain containing 9
View Full Product Details