Product Details

SNP ID
rs2609483
Assay Type
Functionally tested
NCBI dbSNP Submissions
64
Location
Chr.1:181322730 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTATGTTCATGTGTTATTGCTGTGG[A/C]AGATTCAGTCTCTGAATCACAGGGC
Phenotype
MIM: 601013
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CACNA1E PubMed Links

Gene Details

Gene
CACNA1E
Gene Name
calcium voltage-gated channel subunit alpha1 E
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000721.3 Intron NP_000712.2
NM_001205293.1 Intron NP_001192222.1
NM_001205294.1 Intron NP_001192223.1
XM_017002243.1 Intron XP_016857732.1
XM_017002244.1 Intron XP_016857733.1
XM_017002245.1 Intron XP_016857734.1
XM_017002246.1 Intron XP_016857735.1
XM_017002247.1 Intron XP_016857736.1
XM_017002248.1 Intron XP_016857737.1
XM_017002249.1 Intron XP_016857738.1
XM_017002250.1 Intron XP_016857739.1
XM_017002251.1 Intron XP_016857740.1

View Full Product Details