Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001039936.2 | 12 | Intron | NP_001035025.1 | ||
NM_001293069.1 | 12 | UTR 5 | NP_001279998.1 | ||
NM_001293070.1 | 12 | Intron | NP_001279999.1 | ||
NM_001293071.1 | 12 | Intron | NP_001280000.1 | ||
NM_001293072.1 | 12 | Intron | NP_001280001.1 | ||
NM_001293073.1 | 12 | Intron | NP_001280002.1 | ||
NM_001293075.1 | 12 | Intron | NP_001280004.1 | ||
NM_001293076.1 | 12 | Intron | NP_001280005.1 | ||
NM_001293077.1 | 12 | Intron | NP_001280006.1 | ||
NM_001293078.1 | 12 | Intron | NP_001280007.1 | ||
NM_001293079.1 | 12 | Intron | NP_001280008.1 | ||
NM_001293080.1 | 12 | Intron | NP_001280009.1 | ||
NM_001293081.1 | 12 | Intron | NP_001280010.1 | ||
NM_004067.3 | 12 | Intron | NP_004058.1 | ||
XM_011515105.2 | 12 | Intron | XP_011513407.1 | ||
XM_011515106.2 | 12 | Intron | XP_011513408.1 | ||
XM_011515107.2 | 12 | Intron | XP_011513409.1 | ||
XM_017011721.1 | 12 | Intron | XP_016867210.1 | ||
XM_017011722.1 | 12 | Intron | XP_016867211.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_019029.2 | 12 | Intron | NP_061902.2 | ||
NM_031311.3 | 12 | Intron | NP_112601.3 | ||
XM_011515437.1 | 12 | Intron | XP_011513739.1 | ||
XM_017012366.1 | 12 | Intron | XP_016867855.1 |