Product Details

SNP ID
rs2617150
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:10376435 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAAACTAAATGACACATGTAAATAA[C/T]CAATGAATCAAAGAATGCAAAAGAC
Phenotype
MIM: 611817
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KLRC4-KLRK1 PubMed Links

Gene Details

Gene
KLRC4-KLRK1
Gene Name
KLRC4-KLRK1 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199805.1 Intron NP_001186734.1
Gene
KLRK1
Gene Name
killer cell lectin like receptor K1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007360.3 Intron NP_031386.2
Gene
LOC101928100
Gene Name
uncharacterized LOC101928100
There are no transcripts associated with this gene.

View Full Product Details