Product Details
- SNP ID
-
rs2765494
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
44
- Location
-
Chr.1:157828675 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TAACCAACTCATTAATGCTCACTTT[A/G]TTGGGTTTTCTTTCTCTCCTACTAA
- Phenotype
-
MIM: 602592
MIM: 606508
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CD5L
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs78132108] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CD5L
- Gene Name
- CD5 molecule like
- Gene
- FCRL1
- Gene Name
- Fc receptor like 1
There are no transcripts associated with this gene.
View Full Product Details