Product Details

SNP ID
rs2765493
Assay Type
Functionally tested
NCBI dbSNP Submissions
61
Location
Chr.1:157828210 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAGGAGGACAGTGGGACCTCAGAAG[A/G]GAGAACCTCTAGGGTGCATCTAGAA
Phenotype
MIM: 602592 MIM: 606508
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CD5L PubMed Links

Gene Details

Gene
CD5L
Gene Name
CD5 molecule like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005894.2 Intron NP_005885.1
XM_005245602.1 Intron XP_005245659.1
XM_017002806.1 Intron XP_016858295.1
Gene
FCRL1
Gene Name
Fc receptor like 1
There are no transcripts associated with this gene.

View Full Product Details