Product Details
- SNP ID
-
rs2129820
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:57068819 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTCCCCTAACTTTCCATGTGCACAC[A/G]CACGCACACACCCCCCTCACTAAAG
- Phenotype
-
MIM: 612115
MIM: 609869
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ARHGEF3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs80242482] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ARHGEF3
- Gene Name
- Rho guanine nucleotide exchange factor 3
- Gene
- SPATA12
- Gene Name
- spermatogenesis associated 12
View Full Product Details