Product Details

SNP ID
rs2855550
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:30887186 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTGGACTCCTATGTGGGAGGGCA[A/G]CAGGGTGAGCTGGTCCAGGCTTATC
Phenotype
MIM: 600408
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DDR1 PubMed Links

Gene Details

Gene
DDR1
Gene Name
discoidin domain receptor tyrosine kinase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001202521.1 Intron NP_001189450.1
NM_001202522.1 Intron NP_001189451.1
NM_001202523.1 Intron NP_001189452.1
NM_001297652.1 Intron NP_001284581.1
NM_001297653.1 Intron NP_001284582.1
NM_001297654.1 Intron NP_001284583.1
NM_001954.4 Intron NP_001945.3
NM_013993.2 Intron NP_054699.2
NM_013994.2 Intron NP_054700.2
XM_006715185.2 Intron XP_006715248.1
XM_011514882.2 Intron XP_011513184.1
XM_011514883.1 Intron XP_011513185.1
XM_011514884.1 Intron XP_011513186.1
XM_011514885.1 Intron XP_011513187.1
XM_011514886.1 Intron XP_011513188.1
XM_011514887.1 Intron XP_011513189.1
XM_011514888.2 Intron XP_011513190.1
XM_017011268.1 Intron XP_016866757.1
XM_017011269.1 Intron XP_016866758.1
Gene
MIR4640
Gene Name
microRNA 4640
There are no transcripts associated with this gene.

View Full Product Details