Product Details

SNP ID
rs2187614
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:6791091 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGATTTCCCTCTTTCAATGTGGGAG[C/G]ACTTACATAGGAGAGGTCTATATCT
Phenotype
MIM: 186940
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CD4 PubMed Links

Gene Details

Gene
CD4
Gene Name
CD4 molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000616.4 Intron NP_000607.1
NM_001195014.2 Intron NP_001181943.1
NM_001195015.2 Intron NP_001181944.1
NM_001195016.2 Intron NP_001181945.1
NM_001195017.2 Intron NP_001181946.1
XM_017020228.1 Intron XP_016875717.1

View Full Product Details