Product Details

SNP ID
rs2145841
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:32445068 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTGTAGTTTCACTGGAAAGGAGGA[C/G]CCCACCCAAGAGGTGGGGTCGACAT
Phenotype
MIM: 612990
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ASXL1 PubMed Links

Gene Details

Gene
ASXL1
Gene Name
additional sex combs like 1, transcriptional regulator
There are no transcripts associated with this gene.

Gene
NOL4L
Gene Name
nucleolar protein 4 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256798.1 4093 UTR 3 NP_001243727.1
NM_080616.4 4093 UTR 3 NP_542183.2
XM_005260282.4 4093 Intron XP_005260339.1
XM_005260283.4 4093 Intron XP_005260340.1
XM_005260284.4 4093 Intron XP_005260341.1
XM_005260285.4 4093 Intron XP_005260342.2
XM_005260286.4 4093 Intron XP_005260343.1
XM_005260288.2 4093 Intron XP_005260345.1
XM_005260289.4 4093 Intron XP_005260346.1
XM_006723702.3 4093 Intron XP_006723765.1
XM_006723703.3 4093 Intron XP_006723766.1
XM_011528562.2 4093 Intron XP_011526864.1
XM_011528563.1 4093 Intron XP_011526865.1
XM_017027669.1 4093 Intron XP_016883158.1
XM_017027670.1 4093 Intron XP_016883159.1

View Full Product Details