Product Details

SNP ID
rs2848931
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:74256827 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGCAAAGCAGTTATGAGACCCACC[A/T]ACCTTGAAACACCGCCTTTAAGGTT
Phenotype
MIM: 613218
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CYB5A PubMed Links

Gene Details

Gene
CYB5A
Gene Name
cytochrome b5 type A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001190807.2 420 Intron NP_001177736.1
NM_001914.3 420 Intron NP_001905.1
NM_148923.3 420 Intron NP_683725.1
XM_011525835.2 420 UTR 3 XP_011524137.1

View Full Product Details