Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_000368.4 | 5049 | UTR 3 | NP_000359.1 | ||
NM_001162426.1 | 5049 | UTR 3 | NP_001155898.1 | ||
NM_001162427.1 | 5049 | UTR 3 | NP_001155899.1 | ||
XM_005272211.1 | 5049 | UTR 3 | XP_005272268.1 | ||
XM_006717271.1 | 5049 | UTR 3 | XP_006717334.1 | ||
XM_011518979.2 | 5049 | UTR 3 | XP_011517281.1 | ||
XM_017015096.1 | 5049 | UTR 3 | XP_016870585.1 | ||
XM_017015097.1 | 5049 | UTR 3 | XP_016870586.1 | ||
XM_017015098.1 | 5049 | UTR 3 | XP_016870587.1 | ||
XM_017015099.1 | 5049 | UTR 3 | XP_016870588.1 | ||
XM_017015100.1 | 5049 | UTR 3 | XP_016870589.1 | ||
XM_017015101.1 | 5049 | UTR 3 | XP_016870590.1 |