Product Details

SNP ID
rs2253223
Assay Type
Functionally tested
NCBI dbSNP Submissions
11
Location
Chr.1:198523726 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAAGATCATCAATGCCATGAGTGG[A/G]AAGGATGAACTTTCATAAAATATTG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ATP6V1G3 PubMed Links

Gene Details

Gene
ATP6V1G3
Gene Name
ATPase H+ transporting V1 subunit G3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320218.1 Intron NP_001307147.1
NM_133262.2 Intron NP_573569.1
NM_133326.1 Intron NP_579872.1
XM_006711163.3 Intron XP_006711226.1
XM_011509186.2 Intron XP_011507488.1
XM_011509187.2 Intron XP_011507489.1

View Full Product Details