Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001010986.2 | 6238 | UTR 3 | NP_001010986.1 | ||
NM_173694.4 | 6238 | UTR 3 | NP_775965.2 | ||
XM_017029439.1 | 6238 | UTR 3 | XP_016884928.1 | ||
XM_017029440.1 | 6238 | UTR 3 | XP_016884929.1 | ||
XM_017029441.1 | 6238 | UTR 3 | XP_016884930.1 | ||
XM_017029442.1 | 6238 | UTR 3 | XP_016884931.1 | ||
XM_017029443.1 | 6238 | UTR 3 | XP_016884932.1 | ||
XM_017029444.1 | 6238 | UTR 3 | XP_016884933.1 | ||
XM_017029445.1 | 6238 | Intron | XP_016884934.1 | ||
XM_017029446.1 | 6238 | UTR 3 | XP_016884935.1 | ||
XM_017029447.1 | 6238 | UTR 3 | XP_016884936.1 | ||
XM_017029448.1 | 6238 | UTR 3 | XP_016884937.1 | ||
XM_017029449.1 | 6238 | UTR 3 | XP_016884938.1 |