Product Details
- SNP ID
-
rs2850545
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.18:50887865 on Build GRCh38
- Set Membership
-
Validated
- Context Sequence [VIC/FAM]
- TATATCCAGGCACTAAAGACATTAG[A/C]AAAGATCTTTAAAATAAATATTTAA
- Phenotype
-
MIM: 154270
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ME2
PubMed Links
Gene Details
- Gene
- ME2
- Gene Name
- malic enzyme 2
View Full Product Details