Product Details

SNP ID
rs2850765
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:63330216 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATTTTTAGGTCTTCAATTTTTGTA[C/T]AGAGTTTTTATGTTTTTTAAGTTTT
Phenotype
MIM: 136440
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KDSR PubMed Links

Gene Details

Gene
KDSR
Gene Name
3-ketodihydrosphingosine reductase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002035.2 2585 UTR 3 NP_002026.1
XM_005266677.2 2585 UTR 3 XP_005266734.1
XM_017025690.1 2585 UTR 3 XP_016881179.1

View Full Product Details