Product Details

SNP ID
rs2836655
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.21:38744120 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCAAAAAGATCCGTCTTTTCCCCAC[C/T]ACGGTAAAAAAAAACGTAATTCCTT
Phenotype
MIM: 611723
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
LINC00114 PubMed Links
Additional Information
For this assay, SNP(s) [rs145111784] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
LINC00114
Gene Name
long intergenic non-protein coding RNA 114
There are no transcripts associated with this gene.

View Full Product Details