Product Details

SNP ID
rs2075097
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:39866720 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGCCCAGGGAGACATTCCCTACCC[C/G]CTACTCACAAACTTACCCACACACC
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FCGBP PubMed Links

Gene Details

Gene
FCGBP
Gene Name
Fc fragment of IgG binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003890.2 Intron NP_003881.2

View Full Product Details