Product Details

SNP ID
rs2076212
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:43927090 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCACCAGCTCATCTCCGGCAAAATA[G/T]GCATCTCTCTTACCAGAGTGTCTGA
Phenotype
MIM: 609567
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PNPLA3 PubMed Links

Gene Details

Gene
PNPLA3
Gene Name
patatin like phospholipase domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_025225.2 516 Missense Mutation GGC,TGC G115C NP_079501.2

View Full Product Details