Product Details

SNP ID
rs2229182
Assay Type
DME
NCBI dbSNP Submissions
NA
Location
Chr.15:100885290 on Build GRCh38
Set Membership
DME Validated Inventoried
Context Sequence [VIC/FAM]
AGATATTTATCAACAATGAATGGCA[C/G]GAATCCAAGAGTGGGAAAAAGTTTG
Phenotype
MIM: 600463
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ALDH1A3 PubMed Links

Gene Details

Gene
ALDH1A3
Gene Name
aldehyde dehydrogenase 1 family member A3
There are no transcripts associated with this gene.

Gene
LOC101927751
Gene Name
uncharacterized LOC101927751
There are no transcripts associated with this gene.

Gene
LOC105369201
Gene Name
uncharacterized LOC105369201
There are no transcripts associated with this gene.

View Full Product Details