Product Details

SNP ID
rs2241933
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.17:4796652 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CAGCGACGGACAGATTTTGTATTTT[G/T]GGAACTAGGTAGAGAGATCATTGAG
Phenotype
MIM: 600307
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
GLTPD2 PubMed Links

Gene Details

Gene
GLTPD2
Gene Name
glycolipid transfer protein domain containing 2
There are no transcripts associated with this gene.

Gene
LOC107984973
Gene Name
atherin-like
There are no transcripts associated with this gene.

Gene
PSMB6
Gene Name
proteasome subunit beta 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270481.1 Intron NP_001257410.1
NM_002798.2 Intron NP_002789.1

View Full Product Details