Product Details

SNP ID
rs2948818
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:125083154 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGGAGACTTTTCACTCTATATATCT[C/T]TTGGGTGTGGAAACATGTGAGTGTA
Phenotype
MIM: 611316
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SLC12A8 PubMed Links

Gene Details

Gene
SLC12A8
Gene Name
solute carrier family 12 member 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001195483.1 2931 UTR 3 NP_001182412.1
NM_024628.5 2931 UTR 3 NP_078904.3

View Full Product Details