Product Details

SNP ID
rs2239897
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.11:2870735 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GGTCAGGACCCACTGCCGAGCACCA[A/G]CTCCCCATGCCACCACCAGGGGTGC
Phenotype
MIM: 610980
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KCNQ1-AS1 PubMed Links

Gene Details

Gene
KCNQ1-AS1
Gene Name
KCNQ1 antisense RNA 1
There are no transcripts associated with this gene.

Gene
KCNQ1DN
Gene Name
KCNQ1 downstream neighbor (non-protein coding)
There are no transcripts associated with this gene.

View Full Product Details