Product Details

SNP ID
rs2240291
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:96972802 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AACAAAGCACAAGATCAGTGATAAA[A/T]GGCAAATGATGCTTTGCCTCAATGT
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
DLX6-AS1 PubMed Links

Gene Details

Gene
DLX6-AS1
Gene Name
DLX6 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details