Product Details

SNP ID
rs2240451
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:108570599 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTGTCAGTTTTAGATCCGCCCTTTG[A/T]CTAGGGAGCAACCTGCAGTTGACGC
Phenotype
MIM: 602634 MIM: 612534
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
DNAJB9 PubMed Links

Gene Details

Gene
DNAJB9
Gene Name
DnaJ heat shock protein family (Hsp40) member B9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012328.2 Intron NP_036460.1
Gene
THAP5
Gene Name
THAP domain containing 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130475.2 Intron NP_001123947.1
NM_001287598.1 Intron NP_001274527.1
NM_001287599.1 Intron NP_001274528.1
NM_001287601.1 Intron NP_001274530.1
NM_182529.3 Intron NP_872335.2

View Full Product Details