Product Details

SNP ID
rs2273399
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.14:94304653 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GTAGTGAGACCTGCTGTGCGTTCTC[A/G]CTTTCCTGACTCATTGCTGGGACCC
Phenotype
MIM: 122500
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
SERPINA6 PubMed Links

Gene Details

Gene
SERPINA6
Gene Name
serpin family A member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001756.3 Intron NP_001747.2

View Full Product Details