Product Details

SNP ID
rs2293744
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.11:298316 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AACTCTCGGGTTAGGGTGAAGACCC[T/C]GGGGTCTGACTGGTTCAGCCTTAGG
Phenotype
MIM: 605578 MIM: 614757 MIM: 617032
Polymorphism
T/C, Transition Substitution
Allele Nomenclature
Literature Links
IFITM2 PubMed Links
Additional Information
For this assay, SNP(s) [rs149533423] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
IFITM2
Gene Name
interferon induced transmembrane protein 2
There are no transcripts associated with this gene.

Gene
IFITM5
Gene Name
interferon induced transmembrane protein 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001025295.2 620 UTR 3 NP_001020466.1
Gene
PGGHG
Gene Name
protein-glucosylgalactosylhydroxylysine glucosidase
There are no transcripts associated with this gene.

View Full Product Details