Product Details

SNP ID
rs2293809
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:89758087 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGAACGGGGAGGCCATCTGCAGCGC[C/T]CTGCCCACCATTCCCTACCACAAAC
Phenotype
MIM: 603455
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC101929709 PubMed Links

Gene Details

Gene
LOC101929709
Gene Name
uncharacterized LOC101929709
There are no transcripts associated with this gene.

Gene
RIPK2
Gene Name
receptor interacting serine/threonine kinase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003821.5 305 Silent Mutation GCC,GCT A9A NP_003812.1
XM_005251092.3 305 UTR 5 XP_005251149.1
XM_011517357.2 305 Intron XP_011515659.1

View Full Product Details