Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001042413.1 | 5801 | UTR 3 | NP_001035878.1 | ||
NM_152629.3 | 5801 | UTR 3 | NP_689842.3 | ||
XM_005251386.4 | 5801 | UTR 3 | XP_005251443.1 | ||
XM_005251387.4 | 5801 | UTR 3 | XP_005251444.1 | ||
XM_005251388.4 | 5801 | UTR 3 | XP_005251445.1 | ||
XM_005251389.4 | 5801 | Intron | XP_005251446.1 | ||
XM_006716731.3 | 5801 | Intron | XP_006716794.1 | ||
XM_011517763.2 | 5801 | UTR 3 | XP_011516065.1 | ||
XM_011517764.2 | 5801 | UTR 3 | XP_011516066.1 | ||
XM_011517765.2 | 5801 | Intron | XP_011516067.1 | ||
XM_011517766.2 | 5801 | UTR 3 | XP_011516068.1 | ||
XM_011517767.2 | 5801 | UTR 3 | XP_011516069.1 | ||
XM_011517769.2 | 5801 | Intron | XP_011516071.1 | ||
XM_017014361.1 | 5801 | UTR 3 | XP_016869850.1 |