Product Details

SNP ID
rs2298314
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:35089406 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAGGGGCAAACACTTTCCAGACCAT[A/G]AGATGCCTGCGAAGGATGGAGGCTG
Phenotype
MIM: 602956 MIM: 614730
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FANCG PubMed Links

Gene Details

Gene
FANCG
Gene Name
Fanconi anemia complementation group G
There are no transcripts associated with this gene.

Gene
PIGO
Gene Name
phosphatidylinositol glycan anchor biosynthesis class O
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001201484.1 2357 Silent Mutation CTC,CTT L621L NP_001188413.1
NM_032634.3 2357 Silent Mutation CTC,CTT L1038L NP_116023.2
NM_152850.3 2357 Silent Mutation CTC,CTT L621L NP_690577.2
XM_005251619.3 2357 Silent Mutation CTC,CTT L1038L XP_005251676.1
XM_017015222.1 2357 Silent Mutation CTC,CTT L1038L XP_016870711.1
XM_017015223.1 2357 Silent Mutation CTC,CTT L621L XP_016870712.1
XM_017015224.1 2357 Silent Mutation CTC,CTT L621L XP_016870713.1

View Full Product Details