Product Details

SNP ID
rs2995229
Assay Type
Functionally tested
NCBI dbSNP Submissions
20
Location
Chr.1:36307989 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTGAGCACCCATCCAAAGGGTCCCC[C/T]ACTCCCTCAGCCACTCCCAAGGTTG
Phenotype
MIM: 603809
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SH3D21 PubMed Links

Gene Details

Gene
SH3D21
Gene Name
SH3 domain containing 21
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001162530.1 Intron NP_001156002.1
NM_024676.4 Intron NP_078952.4
XM_011542153.2 Intron XP_011540455.1
XM_017002340.1 Intron XP_016857829.1
XM_017002341.1 Intron XP_016857830.1
XM_017002342.1 Intron XP_016857831.1
XM_017002343.1 Intron XP_016857832.1
XM_017002344.1 Intron XP_016857833.1
XM_017002345.1 Intron XP_016857834.1
XM_017002346.1 Intron XP_016857835.1
Gene
THRAP3
Gene Name
thyroid hormone receptor associated protein 3
There are no transcripts associated with this gene.

View Full Product Details