Product Details
- SNP ID
-
rs2306034
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:46857133 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GTGCAATAAGATCCTTCCCCCTTTC[C/T]AATCAAGAAAATGCCACTTTTCTAC
- Phenotype
-
MIM: 604270
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
LRP4
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs147363340] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LRP4
- Gene Name
- LDL receptor related protein 4
- Gene
- LRP4-AS1
- Gene Name
- LRP4 antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details