Product Details

SNP ID
rs2306125
Assay Type
Functionally tested
NCBI dbSNP Submissions
34
Location
Chr.1:155052885 on Build GRCh38
Set Membership
HapMap JSNP
Context Sequence [VIC/FAM]
TGAGGAGCTGGTGGGTAGAGCTCAC[C/T]GTAGTGGGTCTATCCCAGGCCCAGC
Phenotype
MIM: 605548
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ADAM15 PubMed Links

Gene Details

Gene
ADAM15
Gene Name
ADAM metallopeptidase domain 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001261464.1 Intron NP_001248393.1
NM_001261465.1 Intron NP_001248394.1
NM_001261466.1 Intron NP_001248395.1
NM_003815.4 Intron NP_003806.3
NM_207191.2 Intron NP_997074.1
NM_207194.2 Intron NP_997077.1
NM_207195.2 Intron NP_997078.1
NM_207196.2 Intron NP_997079.1
NM_207197.2 Intron NP_997080.1
Gene
DCST1
Gene Name
DC-STAMP domain containing 1
There are no transcripts associated with this gene.

Gene
LOC100505666
Gene Name
uncharacterized LOC100505666
There are no transcripts associated with this gene.

View Full Product Details