Product Details
- SNP ID
-
hCV16201346
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:73143701 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CGAACAGATCCTGTTAGCATAGGGT[A/G]AAAATGGAAACTACTTCTTTTAAAA
- Phenotype
-
MIM: 104311
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PSEN1
PubMed Links
Gene Details
- Gene
- PSEN1
- Gene Name
- presenilin 1
View Full Product Details