Product Details

SNP ID
rs2335772
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:128053693 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGCCCATGTGCTGCTTTTAATGGAC[G/T]AATTTGTAAAGCTTTACCCCCTCAA
Phenotype
MIM: 609213
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
SEC61A1 PubMed Links

Gene Details

Gene
SEC61A1
Gene Name
Sec61 translocon alpha 1 subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013336.3 Intron NP_037468.1

View Full Product Details