Product Details

SNP ID
rs3026051
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:117243977 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTCTGCTCTTATCCCCAGTCCCCA[A/T]CATCCCAAGTTATCTGTACAGCATT
Phenotype
MIM: 193001
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
SLC18A2 PubMed Links

Gene Details

Gene
SLC18A2
Gene Name
solute carrier family 18 member A2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003054.4 291 Missense Mutation AAC,ATC N43I NP_003045.2

View Full Product Details