Product Details
- SNP ID
-
rs2343664
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:136879541 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTCCTCAAAAAGATATTTTGAATTC[G/T]TAATCCCTAGTACTTAAGAATGTGA
- Phenotype
-
MIM: 600508
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
NCK1
PubMed Links
Gene Details
- Gene
- NCK1
- Gene Name
- NCK adaptor protein 1
View Full Product Details