Product Details

SNP ID
rs2426156
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.20:50518762 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACGCCACCTCCCTGTTGCCATAGTG[G/T]TAAGGATCATGCTCCGAGGTGGGGT
Phenotype
MIM: 176885
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
PTPN1 PubMed Links
Additional Information
For this assay, SNP(s) [rs60515027] are located under a probe and SNP(s) [rs35044946] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
PTPN1
Gene Name
protein tyrosine phosphatase, non-receptor type 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278618.1 Intron NP_001265547.1
NM_002827.3 Intron NP_002818.1

View Full Product Details