Product Details
- SNP ID
-
rs2427494
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:63097157 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TGCACTCTAATTCTGCCCCAGGGAA[C/G]CCCAAGGTCAGGATTAGCATCCCCC
- Phenotype
-
MIM: 610556
MIM: 610557
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
HAR1A
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs147467856] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- HAR1A
- Gene Name
- highly accelerated region 1A (non-protein coding)
There are no transcripts associated with this gene.
- Gene
- HAR1B
- Gene Name
- highly accelerated region 1B (non-protein coding)
There are no transcripts associated with this gene.
View Full Product Details