Product Details
- SNP ID
-
rs2531702
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:20019189 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTCATCCTCTCACCTAGCTCCATGC[A/G]GACAGCTGTGAGGCCCCCGGAGAAA
- Phenotype
-
MIM: 602269
MIM: 616830
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ARVCF
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs74731343] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ARVCF
- Gene Name
- armadillo repeat gene deleted in velocardiofacial syndrome
There are no transcripts associated with this gene.
- Gene
- TANGO2
- Gene Name
- transport and golgi organization 2 homolog
View Full Product Details