Product Details
- SNP ID
-
rs2505323
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:27398296 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CAGTCTTCAGCTATGCGTCGTAATT[A/G]GAATAACAATATTTTCTTTTTGGCT
- Phenotype
-
MIM: 611791
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PTCHD3
PubMed Links
Gene Details
- Gene
- PTCHD3
- Gene Name
- patched domain containing 3
There are no transcripts associated with this gene.
View Full Product Details