Product Details

SNP ID
rs2622925
Assay Type
Functionally tested
NCBI dbSNP Submissions
34
Location
Chr.1:47352558 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGTCACAGATTCTCATTACTTCTTT[C/G]ACCAGTTCAAGGGCATAATTGAGTT
Phenotype
MIM: 191710
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CMPK1 PubMed Links

Gene Details

Gene
CMPK1
Gene Name
cytidine/uridine monophosphate kinase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136140.1 Intron NP_001129612.1
NM_016308.2 Intron NP_057392.1

View Full Product Details