Product Details

SNP ID
rs2676401
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:33489910 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAATGCTAAAGATGATGTCAGACTT[C/T]GACAGAAAATTCTTTTTTCTTAGTT
Phenotype
MIM: 614426
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MAK16 PubMed Links

Gene Details

Gene
MAK16
Gene Name
MAK16 homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032509.3 Intron NP_115898.2
Gene
TTI2
Gene Name
TELO2 interacting protein 2
There are no transcripts associated with this gene.

View Full Product Details