Product Details

SNP ID
rs2742971
Assay Type
Functionally tested
NCBI dbSNP Submissions
28
Location
Chr.1:23509226 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGCTCAGCTGGAGATGTGAGGCCA[A/G]GAACAGGGGAAGAGGCAAAAGGGCC
Phenotype
MIM: 600426
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
E2F2 PubMed Links

Gene Details

Gene
E2F2
Gene Name
E2F transcription factor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004091.3 1873 UTR 3 NP_004082.1
XM_005245748.3 1873 UTR 3 XP_005245805.1
XM_005245749.3 1873 UTR 3 XP_005245806.1
XM_011540868.2 1873 UTR 3 XP_011539170.1
XM_011540870.2 1873 Intron XP_011539172.1
XM_011540871.2 1873 UTR 3 XP_011539173.1

View Full Product Details