Product Details

SNP ID
rs35106830
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:161749953 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGGAGCCAGATCACCTGAGGGAAG[A/C]GGGCATCACGGCCGTGCTAACAGTG
Phenotype
MIM: 604835
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
DUSP12 PubMed Links

Gene Details

Gene
DUSP12
Gene Name
dual specificity phosphatase 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007240.2 186 Missense Mutation GAG,GCG E51A NP_009171.1
XM_005244862.2 186 UTR 5 XP_005244919.1

View Full Product Details