Product Details

SNP ID
rs2277767
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:63035585 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTCACCTGTGTCAGGCCTGGGTGCC[A/G]CCAGGAGGCCTCCTTCCAGCTCTCT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LINC00029 PubMed Links

Gene Details

Gene
LINC00029
Gene Name
long intergenic non-protein coding RNA 29
There are no transcripts associated with this gene.

Gene
LINC01056
Gene Name
long intergenic non-protein coding RNA 1056
There are no transcripts associated with this gene.

Gene
LOC63930
Gene Name
uncharacterized LOC63930
There are no transcripts associated with this gene.

View Full Product Details