Product Details

SNP ID
rs6075101
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.20:16749834 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TGATTCTCACTATCAGCAAAGAGGA[C/T]GCCTGCGAGTTTCTCAGAAGACACT
Phenotype
MIM: 606067 MIM: 603520
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OTOR PubMed Links

Gene Details

Gene
OTOR
Gene Name
otoraplin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020157.3 Intron NP_064542.1
XM_017027959.1 Intron XP_016883448.1
Gene
SNRPB2
Gene Name
small nuclear ribonucleoprotein polypeptide B2
There are no transcripts associated with this gene.

View Full Product Details