Product Details

SNP ID
rs45549132
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:49290771 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATCGTCAGGTCTGCAAGGAACCTG[G/T]AGAAGGGCCACCAGAGTGTGGGATG
Phenotype
MIM: 607972
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
SLC6A16 PubMed Links

Gene Details

Gene
SLC6A16
Gene Name
solute carrier family 6 member 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014037.2 Intron NP_054756.2
XM_005258820.2 Intron XP_005258877.1
XM_006723168.3 Intron XP_006723231.1
XM_011526859.2 Intron XP_011525161.2
XM_011526860.2 Intron XP_011525162.1
XM_011526861.2 Intron XP_011525163.1
XM_017026712.1 Intron XP_016882201.1

View Full Product Details